Integrerte vernebriller, Peltor™
Leverandør: 3M
V6B
MMMAV6BEA 420 NOK
MMMAV6B
Integrerte vernebriller, Peltor™
Vernebriller
3M™ Integrated Eyewear V6B have a clear lens and are easy to attach to the headband of 3M™ Safety helmets. The unique design allows the glasses to slide easily between the helmet shell and headband.
  • Glasses easily attach to the headband of 3M™ Safety helmets for eye protection
  • Fit closely to the wearer's cheeks, providing excellent protection from particles and dust
  • Durable lens coating helps minimise physical and chemical damage
  • Easy to adjust for a good fit
Sertifiseringer: EN 166; (EN 172)