Rapid readout biological indicator for ethylene oxide sterilisation, 1294, Attest™
Leverandør: 3M MEDICAL
1294-S
MMME1294-SEA 53275 NOK
MMME1294-S
Rapid readout biological indicator for ethylene oxide sterilisation, 1294, Attest™
Steriliseringsindikatorer
Rapid readout biological indicator for EO (ethylene oxide) sterilization.
  • Self-contained biological indicator design reduces the risk of transfer positives
  • Conforms with ISO 11138:2006 Parts 1 and 2 - Biological Indicators
  • Used in conjunction with the 3M™ Attest™ Auto-reader 390G
  • Used to monitor ethylene oxide (EO) sterilization cycles
  • Four hour readout
  • Green cap
The 3M™ A t t e s t™ 1294-S Rapid Readout Biological Indicator for Ethylene Oxide is a reliable, convenient, and efficient sterilization monitoring indicator designed for validation andMore Product Information