Super Rapid Readout Steam Challenge Pack 1496VX, 3M™ Attest™
Leverandør: 3M MEDICAL
1496V
MMME1496VEA 6825 NOK
MMME1496V
Super Rapid Readout Steam Challenge Pack 1496VX, 3M™ Attest™
Steriliseringsindikatorer
Twenty four minute Biological Indicator Challenge Pack for 132°C (270°F) 4 minute and 135°C (275°F) 3 minute Dynamic Air Removal Steam Sterilization. Each pack contains one 3M™ Attest™ Super Rapid Readout Biological Indicator 1492V.
  • Provides BI results in 24 minutes in 3M Attest 490/490H Auto-readers with software version 4.2.7 or greater
  • Dynamic-air-removal (pre-vacuum and SFPP): 132 ˚C (270˚F) 4 min, 135 ˚C (275˚F) 3 min
  • Contains a 3M™ Attest ™ Super Rapid Readout Biological Indicator 1492V
  • The biological indicator within the challenge pack contains