Dual auto-reader for steam and vaporised hydrogen peroxide 490, Attest™
Leverandør: 3M MEDICAL
490
MMME490EA 0 NOK
MMME490
Dual auto-reader for steam and vaporised hydrogen peroxide 490, Attest™
Steriliseringsindikatorer
Auto-reader for use with 3M™ Attest™ Rapid Readout Biological Indicators and process challenge packs for steam. Includes ethernet cable for web app connectivity.
  • Designed for use with Attest™ Super Rapid Readout Biological Indicators 1491 and 1492
  • Intelligent display with time-to-result countdown feature
  • Display viewable remotely using your Smartphone or PC, manage record-keeping with web app and your internet connected PC
  • Reader will still connect to leading instrument Tracking Software Systems
The 3M™ Attest™ 490 Auto reader for steam reads theMore Product Information