Tegaderm™ Antimicrobial I.V. Advanced Securement Dressing
Leverandør: 3M MEDICAL
9132
MMME9132EA 2200 NOK
MMME9132
Tegaderm™ Antimicrobial I.V. Advanced Securement Dressing
Førstehjelpsutstyr Bandasjer
The Tegaderm™ Antimicrobial I.V. Advanced Securement Dressing is a peripheral IV securement dressing with chlorhexidine gluconate (CHG) formulated into the dressing adhesive. Its integrated design combines antimicrobial protection with site visibility, catheter securement and consistent application for peripheral IVs.
  • Designed to minimise catheter movement and dislodgement
  • Breathable; semi-permeable
  • Comfortable to wear and conformable, permits free movement
  • Large securement tape strip with notch promotes consistent, correct application
  • Transparent film