Rapid readout biological indicator for EO 1294, Attest™
Leverandør: 3M MEDICAL
M1294E
MMMEM1294EEA 6250 NOK
MMMEM1294E
Rapid readout biological indicator for EO 1294, Attest™
Steriliseringsindikatorer
Four-hour, green cap, Rapid Readout Biological Indicator for EO Sterilization . Biological indicators and test packs that monitor the effectiveness of the EO sterilisation process and provide results within four hours.
  • After sterilisation, the vial is 'crushed' to join the growth media with the processed spore strip
  • The BI is incubated for 4 hours for a fluorescent readout
  • Test packs present a challenge to the sterilization process as do the AAMI recommended packs
  • Every load monitoring and quarantining can eliminate recalls
The self-contained BI forMore Product Information