Autoclave test packs, Bowie and Dick type, Comply™
Leverandør: 3M MEDICAL
M1301
MMMEM1301EA 6850 NOK
MMMEM1301
Autoclave test packs, Bowie and Dick type, Comply™
Steriliseringsindikatorer
With unique colour chemistry, the 3M™ Comply™ Disposable Bowie and Dick Type Autoclave Test Pack has been designed for single use in porous load sterilizers for daily Bowie and Dick tests.
  • Easy to detect failures according to ISO 11140 Part 1
  • Easy to interpret fail or pass
  • Easy to locate and retrieve test sheet
  • Easy to open "matchbox" system
  • Paper indicator sheet
Bowie-Dick Test Packs are used to monitor the performance of vacuum-assisted pre-vac steam sterilizers; test packs will reproducibly detect residual air in the sterilizer chamber due to leaks,More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Type 1301
Farge Blue
Fast tid 3,5 mins
Driftstemperatur < 230 °C