Time temperature indicators, MonitorMark™
Leverandør: 3M Food Safety
9860H 9860C 9860D 9861A 9860B
MMMP9860HEA 23000 NOK
MMMP9860H MMMP9860C MMMP9860D MMMP9861A MMMP9860B
Time temperature indicators, MonitorMark™
Temperatur indikatorstrips
3M™ MonitorMark™ time temperature indicator solves the challenge of monitoring thermal exposure of temperature sensitive products during transportation and storage.
  • Inexpensive solution for monitoring product exposure to a range of temperatures for a specific period of time
  • Self-adhesive backing for easy attachment to secondary packaging
  • Easily-interpreted visual results
  • Results indicate both exposure and relative time over which exposure occurred
  • Removable film activation strip starts the monitoring process on demand for accuracy
These indicators areMore Product Information
Bestill nå
SPESIFIKASJONER
Test parameters Environmental test: time, temperature
Environmentally Preferable
Lagringstemp 22 °C (72 °F) or less, 20 - 60% relative humidity