Dry-sponge
Leverandør: 3M Food Safety
BP133ES
MMMPBP133ESEA 2400 NOK
MMMPBP133ES
Dry-sponge
Svabere Svabere for prøvetaking
The 3M™ Dry-Sponge is a biocide-free cellulose sponge that comes with a printed sample bag.
  • 1,5×3 inch dry sponge can be used with a wide range buffers and enrichment media and in dry-only environments
  • Simplified packaging convenience minimises handling and preparation and reduces waste
  • Biocide-free sponge does not affect organism viability
  • 5×9 inch printable sample bag included for safe, convenient transportation to lab
  • Decontaminated through gamma irradiation to maintain aseptic conditions
Cellulose is an ideal sampling material for monitoring forMore Product Information