Dry-sponge with glove
Leverandør: 3M Food Safety
BP237SPG
MMMPBP237SPGEA 3150 NOK
MMMPBP237SPG
Dry-sponge with glove
Svabere Svabere for prøvetaking
3M™ Dry-sponge with gloves is a biocide-free cellulose sponge that includes two polyethylene gloves and a sample bag.
  • 1,5×3 inch dry sponge can be used with a wide range buffers and enrichment media and in dry-only environments
  • Simplified packaging convenience, including sponge, sample bag and gloves, minimises handling and preparation and reduces waste
  • 5×9 inch sample bag with 18 oz./0,53 L capacity and included for safe handling and convenient transportation to lab
  • Biocide-free sponge does not affect organism viability
  • Decontaminated through gamma