Enviro swab
Leverandør: 3M Food Safety
ENVSWB100
MMMPENVSWB100EA 4175 NOK
MMMPENVSWB100
Enviro swab
Svabere Svabere for prøvetaking
The 3M™ Enviro Swab is a pre-moistened swab, ready to use for collecting environmental samples on all types of food contact surfaces in the food industry.
  • Ready-to-use swab reaches into corners and hard-to-reach areas for increased accuracy
  • Tamper-evident band indicates if the swab has been used
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant
  • Resealable tube provides tight closure for sample transportation
  • Self-contained swab and vial for increased productivity and efficiency
Each swab is in a close sealedMore Product Information