Molecular detection assay 2 - Campylobacter
Leverandør: 3M Food Safety
MDA2CAM96
MMMPMDA2CAM96EA 12275 NOK
MMMPMDA2CAM96
Molecular detection assay 2 - Campylobacter
Overvåkningssystemer Hygieneovervåkningssystemer
3M™ Molecular Detection Assay 2 - Campylobacter is used with the 3M™ Molecular Detection System for the rapid and specific detection of Campylobacter in enriched food and environmental samples.
  • One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees
  • Use with CE250 3M™ Campylobacter enrichment broth for fast and reliable results
  • Ready-to-use and pre-dispensed reagents provide convenience and minimise the chance of human error
  • Color-coded assay tubes by organism and matching software screens allow for