Molecular detection assay E. coli O157
Leverandør: 3M Food Safety
MDA2ECO96
MMMPMDA2ECO96EA 12100 NOK
MMMPMDA2ECO96
Molecular detection assay E. coli O157
Overvåkningssystemer Hygieneovervåkningssystemer
3M™ Molecular Detection Assay 2 - E. coli O157 (including H7) is used with the 3M™ Molecular Detection System for the rapid and specific pathogen detection of E. coli O157, including H7, in enriched food samples.
  • One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees
  • Ready-to-use and pre-dispensed reagents provide convenience and minimise the chance of human error
  • Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification
  • The ability to run