Molecular detection assay, Salmonella
Leverandør: 3M Food Safety
MDA2SAL96
MMMPMDA2SAL96EA 10675 NOK
MMMPMDA2SAL96
Molecular detection assay, Salmonella
Overvåkningssystemer Hygieneovervåkningssystemer
3M™ Molecular Detection Assay 2 - Salmonella is used with the 3M™ Molecular Detection System for the rapid and specific detection of Salmonella species in enriched food and environmental samples.
  • One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees
  • Ready-to-use and pre-dispensed reagents provide convenience and minimise the chance of human error
  • Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification
  • The ability to run all assays