Blood lancets, VITREX® STERIHEEL® Baby II
Leverandør: Vitrex Medical A/S
331213 332213 333213 334213 335213
MODU331213EA 1900 NOK
MODU331213 MODU332213 MODU333213 MODU334213 MODU335213
Blood lancets, VITREX® STERIHEEL® Baby II
Blodlansetter
For high volume blood flow with minimum depth of penetration.
  • User safety - blade is fully shielded before and after use; activation button secured by safety catch
  • User convenience - pre-loaded, button activated design
  • Patient safety - single-use only
  • Sterilised by gamma irradiation
  • Patient comfort - ultra sharp blade with high speed action securing minimal trauma and fast healing
Application: Primarily newborns and preemies. Can also be recommended for other applications where short penetration depth and/or minimal compression of tissue is important.More Product Information