Manual pulse counter
Leverandør: MOINEAU
1023MW
MOIN1023MWEA 325 NOK
MOIN1023MW
Manual pulse counter
Celletellere Håndteller
Manual mechanical counter with four digit key in a robust metal housing.
  • General reset by the rotary knob side
  • Adding one each finger pressure
  • Folding ring to attach it to your finger