Digital stopwatch, large display
Leverandør: MOINEAU
376CE
MOIN376CEEA 525 NOK
MOIN376CE
Digital stopwatch, large display
Tidsur Stoppeklokker, digitale
Ergonomic stopwatch with large, one-line LCD display.
  • Timing capacity: 24 hours in 1/100 seconds
  • Functions: Split time, clock, date and alarm
  • Water resistant
Weight: 90 g
Leveringsinformasjon: Supplied with 357 battery.