Disposable earplugs, MelLows®
Leverandør: Moldex
7625 7650 7600
MOLD7625EA 1025 NOK
MOLD7625 MOLD7650 MOLD7600
Disposable earplugs, MelLows®
Ørepropper Ørepropper til engangsbruk
Earplugs made from soft PU-foam for protection against lower noise levels.
SNR: 22 dB
Sertifiseringer: EN 352-2:2002
Bestillingsinformasjon: Also available in the hygienic dispenser system MoldexStation.
Forpakning: In pairs.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Safer Human and Environmental Health