Gentamicin sulphate

Leverandør: Molekula
Danger

10336465-5G
MOLE10336465-5GEA 1000 NOK
MOLE10336465-5G
Gentamicin sulphate
Gentamicin sulphate
Smeltepunkt: 218…237 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00270181
CAS-nummer: 1405-41-0
EINECS: 215-778-9
Merck Indeks: 13,04403

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR