(1R)-(+)-Camphanic chloride

Leverandør: Molekula

10505203-1G
MOLE10505203-1GEA 0 NOK
MOLE10505203-1G
(1R)-(+)-Camphanic chloride
(1R)-(+)-Camphanic chloride
Formel: C₁₀H₁₃ClO₃
CAS-nummer: 104530-16-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR