Syklopropylamin

Leverandør: Molekula

Synonymer: Aminocyclopropane

10907539-25G
MOLE10907539-25GEA 1025 NOK
MOLE10907539-25G
Syklopropylamin
Syklopropylamin
Formel: C₃H₇N
MW: 57.1 g/mol
Kokepunkt: 49
Smeltepunkt: –10 °C
Tetthet: 0.8131 g/cm³ (20 °C)
Flash punkt: 1
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001301
CAS-nummer: 765-30-0
EINECS: 212-142-2
FN: 2733
ADR: 3 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR