2-Naphthylacetonitrile

Leverandør: Molekula

12315779-5G
MOLE12315779-5GEA 0 NOK
MOLE12315779-5G
2-Naphthylacetonitrile
2-Naphthylacetonitrile
Formel: C₁₂H₉N
MW: 167,21 g/mol
MDL nummer: MFCD00004122
CAS-nummer: 7498-57-9
EINECS: 231-352-5
FN: 3439
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR