Indole-4-carboxylic acid

Leverandør: Molekula
Warning

15426468-1G
MOLE15426468-1GEA 4125 NOK
MOLE15426468-1G
Indole-4-carboxylic acid
Indole-4-carboxylic acid
Formel: C₉H₇NO₂
MW: 161.16 g/mol
MDL nummer: MFCD00009739
CAS-nummer: 2124-55-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR