Dibenzyl diisopropylphosphoramidite

Leverandør: Molekula

17862617-5G
MOLE17862617-5GEA 2075 NOK
MOLE17862617-5G
Dibenzyl diisopropylphosphoramidite
Dibenzyl diisopropylphosphoramidite
Formel: C₂₀H₂₈NO₂P
MDL nummer: MFCD00191988
CAS-nummer: 108549-23-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR