(S)-(+)-2-Amino-3-methyl-1-butanol

Leverandør: Molekula

Synonymer: L(+)-2-Amino-3-methyl-1-butanol, (S)-2-Amino-3-methylbutan-1-ol, (S)-2-Amino-3-methyl-butan-1-ol, L-Valinol, L(+)-Valinol

19625420-1G
MOLE19625420-1GEA 0 NOK
MOLE19625420-1G
(S)-(+)-2-Amino-3-methyl-1-butanol
(S)-(+)-2-Amino-3-methyl-1-butanol
Formel: C₅H₁₃NO
MW: 103,16 g/mol
Kokepunkt: 187 °C (1013 hPa)
Smeltepunkt: 31…32 °C
Tetthet: 0,92 g/cm³ (20 °C)
MDL nummer: MFCD00064296
CAS-nummer: 2026-48-4
EINECS: 217-975-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR