2'-Deoxycytidine-5'-triphosphoric acid disodium salt (dCTP-Na2)

Leverandør: Molekula

Synonymer: 2'-Deoxycytidine-5'-triphosphoric acid disodium salt, 2′-Deoxycytidine-5′-Triphosphate Trisodium Salt, 2'-Deoxycytidine-5'-triphosphate disodium salt

20106938-100MG
MOLE20106938-100MGEA 0 NOK
MOLE20106938-100MG
2'-Deoxycytidine-5'-triphosphoric acid disodium salt (dCTP-Na2)
2'-Deoxycytidine-5'-triphosphoric acid disodium salt (dCTP-Na2)
Formel: C₉H₁₄N₃Na₂O₁₃P₃
MDL nummer: MFCD00084683
CAS-nummer: 102783-51-7
EINECS: 252-849-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR