Seleninyl dichloride

Leverandør: Molekula
Danger

Synonymer: Selenium dichloride oxide, Selenium oxychloride

23618910-5G
MOLE23618910-5GEA 0 NOK
MOLE23618910-5G
Seleninyl dichloride
Seleninyl dichloride
Formel: Cl₂OSe
MW: 165,86 g/mol
Kokepunkt: 178 °C (1013 hPa)
Smeltepunkt: 8,5 °C
Tetthet: 2,44 g/cm³ (20 °C)
MDL nummer: MFCD00016327
CAS-nummer: 7791-23-3
EINECS: 232-244-0
FN: 2879
ADR: 8,I
Merck Indeks: 12,08578

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR