Metylgult

Leverandør: Molekula
Danger

Synonymer: Dimethyl yellow, Sudan Yellow, 4-(Dimethylamino)azobenzene, 4-(N,N-Dimethylamino)azobenzene

27793472-25G
MOLE27793472-25GEA 1025 NOK
MOLE27793472-25G
Metylgult
Metylgult
Formel: C₁₄H₁₅N₃
MW: 225,29 g/mol
Kokepunkt: 371 °C (1013 hPa)
Smeltepunkt: 113…117 °C
Tetthet: 1,2233 g/cm³ (20 °C)
MDL nummer: MFCD00008308
CAS-nummer: 60-11-7
EINECS: 200-455-7
FN: 2811
ADR: 6.1,III
Merck Indeks: 12,03279

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR