Quinoxaline

Leverandør: Molekula

28973649-100G
MOLE28973649-100GEA 0 NOK
MOLE28973649-100G
Quinoxaline
Quinoxaline
Formel: C₈H₆N₂
MW: 130,15 g/mol
Kokepunkt: 230 °C (1013 hPa)
Smeltepunkt: 30…32 °C
Tetthet: 1,1485 g/cm³ (32 °C)
MDL nummer: MFCD00006719
CAS-nummer: 91-19-0
EINECS: 202-047-4
Merck Indeks: 12,08263

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR