2-Mercapto-5-methyl-1,3,4-thiadiazole

Leverandør: Molekula

29054861-100G
MOLE29054861-100GEA 0 NOK
MOLE29054861-100G
2-Mercapto-5-methyl-1,3,4-thiadiazole
2-Mercapto-5-methyl-1,3,4-thiadiazole
Formel: C₃H₄N₂S₂
MDL nummer: MFCD00038349
CAS-nummer: 29490-19-5
EINECS: 249-667-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR