4-Isopropylbensaldehyd

Leverandør: Molekula
Warning

Synonymer: Cuminaldehyde

30191840-100G
MOLE30191840-100GEA 0 NOK
MOLE30191840-100G
4-Isopropylbensaldehyd
4-Isopropylbensaldehyd
Formel: C₁₀H₁₂O
MW: 148,2 g/mol
Kokepunkt: 235 °C (1013 hPa)
Tetthet: 0,98 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006953
CAS-nummer: 122-03-2
EINECS: 204-516-9
Merck Indeks: 13,02648

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR