Spermidine trihydrochloride

Leverandør: Molekula

31867014-5G
MOLE31867014-5GEA 0 NOK
MOLE31867014-5G
Spermidine trihydrochloride
Spermidine trihydrochloride
Formel: C₇H₂₂Cl₃N₃
Smeltepunkt: 256…258 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00012918
CAS-nummer: 334-50-9
EINECS: 206-379-0
Merck Indeks: 12,08893

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR