Propyl 4-hydroksybenzoat

Leverandør: Molekula
Warning

Synonymer: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester

32467626-1KG
MOLE32467626-1KGEA 2100 NOK
MOLE32467626-1KG
Propyl 4-hydroksybenzoat
Propyl 4-hydroksybenzoat
Formel: C₁₀H₁₂O₃
MW: 180,2 g/mol
Kokepunkt: 292 °C (1013 hPa)
Smeltepunkt: 95 °C
Tetthet: 1,28 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002354
CAS-nummer: 94-13-3
EINECS: 202-307-7
Merck Indeks: 13,07958

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR