Diallyl disulphide

Leverandør: Molekula
Warning

Synonymer: Diallyldisulphide, Allyl disulphide

33421999-100G
MOLE33421999-100GEA 0 NOK
MOLE33421999-100G
Diallyl disulphide
Diallyl disulphide
Formel: C₆H₁₀S₂
MW: 146,28 g/mol
Kokepunkt: 198 °C (1013 hPa)
Tetthet: 1,0237 g/cm³ (15 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00008656
CAS-nummer: 2179-57-9
EINECS: 218-548-6
FN: 2810
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR