2-Metoksybensylamin

Leverandør: Molekula
Danger

Synonymer: (2-Methoxyphenyl)methanamine, 2-(Aminomethyl)anisole, o-Anisylamine

35102525-25G
MOLE35102525-25GEA 0 NOK
MOLE35102525-25G
2-Metoksybensylamin
2-Metoksybensylamin
Formel: C₈H₁₁NO
MW: 137,18 g/mol
Kokepunkt: 227 °C (1013 hPa)
Tetthet: 1,05 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008110
CAS-nummer: 6850-57-3
EINECS: 229-941-7
FN: 2735
ADR: 8,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR