DL-Lithium lactate

Leverandør: Molekula

Synonymer: DL-Lactic acid lithium salt, (±)-2-Hydroxypropanoic acid lithium salt, Lithium 2-hydroxypropanoate, (±)-2-Hydroxypropionic acid lithium salt, (±)-Lactic acid lithium salt

43935171-100G
MOLE43935171-100GEA 1650 NOK
MOLE43935171-100G
DL-Lithium lactate
DL-Lithium lactate
Formel: H₃CCH(OH)COOLi
MW: 96.01 g/mol
Smeltepunkt: 300
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00065399
CAS-nummer: 867-55-0
EINECS: 212-761-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR