Brilliant cresyl blue

Leverandør: Molekula

Synonymer: CI 51010, 1,3-Diamino-7-(diethylamino)-3-methylphenoxazin-5-ium tetrachlorozincate (2:1), Brilliant cresyl blue zinc chloride double salt, Brilliant Blue C, BCB

45545810
MOLE45545810EA 1975 NOK
MOLE45545810
Brilliant cresyl blue
Brilliant cresyl blue
Formel: C₃₄H₄₂Cl₄N₈O₂Zn
MDL nummer: MFCD00148901
CAS-nummer: 81029-05-2
EINECS: 279-675-0

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR