Kaliumtetrahydroborat

Leverandør: Molekula
Danger

53642148-100G
MOLE53642148-100GEA 2700 NOK
MOLE53642148-100G
Kaliumtetrahydroborat
Potassium tetrahydroborate
Formel: H₄BK
Smeltepunkt: 500 °C
Tetthet: 1,17 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011396
CAS-nummer: 13762-51-1
EINECS: 237-360-5
FN: 1870
ADR: 4.3,I
Merck Indeks: 12,07777

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR