Wright's stain for mikroskopifarging

Leverandør: Molekula
Warning

57163864-25G
MOLE57163864-25GEA 1025 NOK
MOLE57163864-25G
Wright's stain for mikroskopifarging
Wright's stain
Tetthet: 0,8 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00082143
CAS-nummer: 68988-92-1
EINECS: 273-541-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR