1,10-Phenanthroline

Leverandør: Molekula
Danger

Synonymer: o-Phenanthroline

62000053-25G
MOLE62000053-25GEA 1425 NOK
MOLE62000053-25G
1,10-Phenanthroline
1,10-Phenanthroline
Formel: C₁₂H₈N₂
MW: 180,21 g/mol
Kokepunkt: 360 °C (1013 hPa)
Smeltepunkt: 117…119 °C
MDL nummer: MFCD00011678
CAS-nummer: 66-71-7
EINECS: 200-629-2
FN: 2811
ADR: 6.1,III
Merck Indeks: 12,07356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5092a8ec81301ed6ed30f75429f9f27784db47a5&vid=106c4325bb7c310de902d3118cf4b31bcee3dc92&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR