1-(2-Pyridylazo)-2-naphthol (PAN)

Leverandør: Molekula

Synonymer: PAN, 1-(2-Pyridylazo)-2-naphthol

63973572-5G
MOLE63973572-5GEA 0 NOK
MOLE63973572-5G
1-(2-Pyridylazo)-2-naphthol (PAN)
1-(2-Pyridylazo)-2-naphthol
Formel: C₁₅H₁₁N₃O
MW: 249,27 g/mol
Kokepunkt: 412 °C (1013 hPa)
Smeltepunkt: 137…140 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004071
CAS-nummer: 85-85-8
EINECS: 201-637-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR