2-Mercapto-5-methoxybenzimidazole

Leverandør: Molekula

66787209-25G
MOLE66787209-25GEA 0 NOK
MOLE66787209-25G
2-Mercapto-5-methoxybenzimidazole
2-Mercapto-5-methoxybenzimidazole
Formel: C₈H₈N₂OS
Kokepunkt: 400 °C (1013 hPa)
Smeltepunkt: 261…263 °C
MDL nummer: MFCD00134581
CAS-nummer: 37052-78-1
EINECS: 253-326-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR