(1S)-(-)-Camphanic acid

Leverandør: Molekula

Synonymer: (1S,4R)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carboxylic acid

71783403-5G
MOLE71783403-5GEA 2225 NOK
MOLE71783403-5G
(1S)-(-)-Camphanic acid
(1S)-(-)-Camphanic acid
Formel: C₁₀H₁₄O₄
MW: 198,22 g/mol
MDL nummer: MFCD00044948
CAS-nummer: 13429-83-9
EINECS: 236-552-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR