Tetrabutylammonium dihydrogen phosphate

Leverandør: Molekula

75044950-5G
MOLE75044950-5GEA 1850 NOK
MOLE75044950-5G
Tetrabutylammonium dihydrogen phosphate
Tetrabutylammonium dihydrogen phosphate
Formel: C₁₆H₃₈NO₄P
MDL nummer: MFCD00064526
CAS-nummer: 5574-97-0
EINECS: 226-947-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR