Gentamicin sulphate

Leverandør: Molekula
Danger

M17289509
MOLEM17289509EA 3000 NOK
MOLEM17289509
Gentamicin sulphate
Gentamicin sulphate
Smeltepunkt: 218…237 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00270181
CAS-nummer: 1405-41-0
EINECS: 215-778-9
Merck Indeks: 13,04403

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR