2-Chloro-p-toluidine

Leverandør: Molekula
Warning

Synonymer: 2-Chloro-4-methylaniline, 4-Amino-3-chlorotoluene

M17655382
MOLEM17655382EA 1975 NOK
MOLEM17655382
2-Chloro-p-toluidine
2-Chloro-p-toluidine
Formel: C₇H₈ClN
Kokepunkt: 224 °C (1013 hPa)
Smeltepunkt: 7 °C
Tetthet: 1,151 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007666
CAS-nummer: 615-65-6
EINECS: 210-440-7
FN: 3429
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR