Guanidiniumtiocyanat

Leverandør: Molekula
Warning

Synonymer: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

M18449163
MOLEM18449163EA 945 NOK
MOLEM18449163
Guanidiniumtiocyanat
Guanidiniumtiocyanat
Formel: CH₅N₃·CHNS
MW: 118.16 g/mol
Smeltepunkt: 118
Tetthet: 1.29 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013027
CAS-nummer: 593-84-0
EINECS: 209-812-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR