Pivalaldehyde (trimethylacetaldehyde)

Leverandør: Molekula

Synonymer: Pivalaldehyde, Pivaldehyde, Trimethylacetaldehyde, 2,2-Dimethylpropionaldehyde, 2,2-Dimethylpropanal

M21845337
MOLEM21845337EA 5500 NOK
MOLEM21845337
Pivalaldehyde (trimethylacetaldehyde)
Pivalaldehyde (trimethylacetaldehyde)
Formel: C₅H₁₀O
MW: 86.13 g/mol
Kokepunkt: 74
Smeltepunkt: 6
Tetthet: 0.771 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00006962
CAS-nummer: 630-19-3
EINECS: 211-134-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR