Diethyl (cyanomethyl)phosphonate

Leverandør: Molekula

M27888340
MOLEM27888340EA 1675 NOK
MOLEM27888340
Diethyl (cyanomethyl)phosphonate
Diethyl (cyanomethyl)phosphonate
Formel: C₆H₁₂NO₃P
MDL nummer: MFCD00001893
CAS-nummer: 2537-48-6
EINECS: 219-806-0
FN: 3278
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR