1-Naphthylacetonitrile

Leverandør: Molekula

Synonymer: 2-(Naphthalen-1-yl)acetonitrile

M28726321
MOLEM28726321EA 835 NOK
MOLEM28726321
1-Naphthylacetonitrile
1-Naphthylacetonitrile
Formel: C₁₂H₉N
MW: 167,21 g/mol
MDL nummer: MFCD00004041
CAS-nummer: 132-75-2
EINECS: 205-078-1
FN: 3439
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR